CAS RN 20367-38-8
CAS RN 20367-38-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Methyl (5-Chloro-1H-benzimidazol-2-yl)carbamate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Albendazole Impurity G, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Albendazole Impurity G, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Albendazole Impurity G, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.