accessibility menu, dialog, popup

UserName

FFNOOOOH

CAS RN 20372-63-8

IUPAC Name: 4,5-difluoro-2-nitrobenzoic acid
Synonyms: 4,5-difluoro-2-nitrobenzoic acid
Molecular Weight (g/mol): 203.1
Molecular Formula: C7H3F2NO4
InChi Key: HGGRAOYTQNFGGN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=C(F)C=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results