accessibility menu, dialog, popup

UserName

CAS RN 204707-42-6

H3COFOOCH3

CAS RN 204707-42-6

IUPAC Name: methyl 4-fluoro-2-methoxybenzoate
Synonyms: methyl 4-fluoro-2-methoxybenzoate
Molecular Weight (g/mol): 184.17
Molecular Formula: C9H9FO3
InChi Key: LJUAEPNTXDJBRX-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(OC)C=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results