accessibility menu, dialog, popup

UserName

CAS RN 2051-85-6

OHONNH(E)

CAS RN 2051-85-6

IUPAC Name: (4E)-3-hydroxy-4-(2-phenylhydrazin-1-ylidene)cyclohexa-2,5-dien-1-one
Synonyms: (4E)-3-hydroxy-4-(2-phenylhydrazin-1-ylidene)cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: PULNESMFDLBKAZ-SDNWHVSQSA-N
SMILES: OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)