accessibility menu, dialog, popup

UserName

CAS RN 205319-10-4

OPPH3CH3CPdClClPhPhPhPh

CAS RN 205319-10-4

IUPAC Name: palladium(2+) [5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphane dichloride
Synonyms: palladium(2+) xantphos dichloride
Molecular Weight (g/mol): 755.95
Molecular Formula: C39H32Cl2OP2Pd
InChi Key: HEYONDYPXIUDCK-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd++].CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results