CAS RN 205526-29-0
IUPAC Name:
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-iodophenyl)propanoic acid
Synonyms:
2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-iodophenyl)propanoic acid
Molecular Weight (g/mol):
513.33
Molecular Formula:
C24H20INO4
InChi Key:
LXOXXTQKKRJNNB-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
N-Fmoc-4-iodo-D-phenylalanine, 98%
CAS: 205526-29-0 Molecular Formula: C24H20INO4 Molecular Weight (g/mol): 513.33 MDL Number: MFCD00672557 InChI Key: LXOXXTQKKRJNNB-UHFFFAOYNA-N Synonym: fmoc-d-phe 4-i-oh,fmoc-4-iodo-d-phenylalanine,fmoc-d-4-iodophenylalanine,fmoc-d-4-iodophe,fmoc-4'-iodo-d-phe,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid,ambotzfaa1584,fmoc-4-iodo-d-phe-oh,fmoc-d-phe 4-i-oh hplc PubChem CID: 16213204 SMILES: OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12