accessibility menu, dialog, popup

UserName

H3C(R)OH(S)HOOO

CAS RN 205639-93-6

IUPAC Name: (5S,6R)-5,6-dihydroxy-4-pentylcyclohexa-1,3-diene-1-carboxylate
Synonyms: (5S,6R)-5,6-dihydroxy-4-pentylcyclohexa-1,3-diene-1-carboxylate
Molecular Weight (g/mol): 225.27
Molecular Formula: C12H17O4
InChi Key: HDNLKEAXAZXFKE-WDEREUQCSA-M
SMILES: CCCCCC1=CC=C([C@@H](O)[C@H]1O)C([O-])=O
Grade 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)