CAS RN 20577-61-1
CAS RN 20577-61-1
IUPAC Name:
methyl 2,4-dioxopentanoate
Synonyms:
methyl 2,4-dioxopentanoate
Molecular Weight (g/mol):
144.13
Molecular Formula:
C6H8O4
InChi Key:
OMHOEQINEXASKE-UHFFFAOYSA-N
SMILES:
COC(=O)C(=O)CC(C)=O
1
–
1
of
1
results
1
Methyl acetopyruvate, 98%, Thermo Scientific™
CAS: 20577-61-1 Molecular Formula: C6H8O4 Molecular Weight (g/mol): 144.126 MDL Number: MFCD00043638 InChI Key: OMHOEQINEXASKE-UHFFFAOYSA-N Synonym: methyl acetopyruvate,methyl acetylpyruvate,pentanoic acid, 2,4-dioxo-, methyl ester,2,4-dioxopentanoic acid methyl,2,4-dioxo-pentanoic acid methyl ester,pentanoic acid,2,4-dioxo-,methyl ester PubChem CID: 88600 IUPAC Name: methyl 2,4-dioxopentanoate SMILES: CC(=O)CC(=O)C(=O)OC