accessibility menu, dialog, popup

UserName

HOOBr

CAS RN 2067-33-6

IUPAC Name: 5-bromopentanoic acid
Synonyms: 5-bromopentanoic acid
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: WNXNUPJZWYOKMW-UHFFFAOYSA-N
SMILES: OC(=O)CCCCBr
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)