accessibility menu, dialog, popup

UserName

CAS RN 206752-32-1

NHSOOOHOHSOOOONa(E/Z)

CAS RN 206752-32-1

IUPAC Name: sodium 4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-7-sulfonaphthalene-2-sulfonate hydrate
Synonyms: sodium 4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-7-sulfonaphthalene-2-sulfonate hydrate
Molecular Weight (g/mol): 463.41
Molecular Formula: C17H14NNaO9S2
InChi Key: BEDIAJJRAYEWBI-UHFFFAOYSA-M
SMILES: O.[Na+].OC1=CC(=CC2=CC(=CC(NC=C3C=CC=CC3=O)=C12)S(O)(=O)=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results