accessibility menu, dialog, popup

UserName

CH3OHOCH3OHONHNNNNH2NNH2HNClNH2NH2Cl(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 206986-79-0

IUPAC Name: N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide bis(acetic acid) hydrate
Synonyms: bis(acetic acid) chlorhexidine hydrate
Molecular Weight (g/mol): 643.57
Molecular Formula: C26H40Cl2N10O5
InChi Key: JGFZDLOXFRFUTG-UHFFFAOYSA-N
SMILES: O.CC(O)=O.CC(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results