accessibility menu, dialog, popup

UserName

CAS RN 206986-81-4

FClHOF

CAS RN 206986-81-4

IUPAC Name: 2-chloro-3,5-difluorophenol
Synonyms: 2-chloro-3,5-difluorophenol
Molecular Weight (g/mol): 164.54
Molecular Formula: C6H3ClF2O
InChi Key: ZQWULOHSTAQTKJ-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC(F)=C1Cl
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results