accessibility menu, dialog, popup

UserName

NNONOOPFFFFFF

CAS RN 207683-26-9

IUPAC Name: [bis(pyrrolidin-1-yl)methylidene](2,5-dioxopyrrolidin-1-yl)oxidanium
Synonyms: [bis(pyrrolidin-1-yl)methylidene](2,5-dioxopyrrolidin-1-yl)oxidanium
Molecular Weight (g/mol): 266.32
Molecular Formula: C13H20N3O3
InChi Key: IFYOZCQVRXPXHU-UHFFFAOYSA-N
SMILES: O=C1CCC(=O)N1[O+]=C(N1CCCC1)N1CCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)