CAS RN 2081-08-5
IUPAC Name:
4-[1-(4-hydroxyphenyl)ethyl]phenol
Synonyms:
1,1-bis(4-hydroxyphenyl)ethane bisphenol E
Molecular Weight (g/mol):
214.26
Molecular Formula:
C14H14O2
InChi Key:
HCNHNBLSNVSJTJ-UHFFFAOYSA-N
SMILES:
CC(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
1
–
2
of
2
results
1
Bisphenol E 100 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Bisphenol E, TRC
CAS: 2081-08-5 Molecular Formula: C14 H14 O2 Molecular Weight (g/mol): 214.26 Synonym: Phenol, 4,4'-ethylidenedi- (6CI,7CI,8CI),4,4'-Ethylidenebis[phenol],1,1-Bis(4-hydroxyphenyl)ethane,4,4'-Ethylidenediphenol,Bisphenol AD,Bisphenol E IUPAC Name: 4-[1-(4-hydroxyphenyl)ethyl]phenol SMILES: CC(c1ccc(O)cc1)c2ccc(O)cc2