CAS RN 2084-18-6
CAS RN 2084-18-6
IUPAC Name:
3-methylbutane-2-thiol
Synonyms:
3-methylbutane-2-thiol
Molecular Weight (g/mol):
104.21
Molecular Formula:
C5H12S
InChi Key:
BFLXFRNPNMTTAA-UHFFFAOYNA-N
SMILES:
CC(C)C(C)S
1
–
1
of
1
results
1
3-Methyl-2-butanethiol, 95%, remainder mainly 2-methyl-1-butanethiol, Thermo Scientific™
CAS: 2084-18-6 Molecular Formula: C5H12S Molecular Weight (g/mol): 104.21 MDL Number: MFCD00039648 InChI Key: BFLXFRNPNMTTAA-UHFFFAOYNA-N Synonym: 3-methyl-2-butanethiol,2-butanethiol, 3-methyl,3-methyl-2-butylthiol,fema no. 3304,3-methyl-2-butane thiol,acmc-209ff6,3-methyl-2-butyl mercaptan,ksc202s8n,1,2-dimethylpropyl hydrosulfide,1,2-dimethylpropyl hydrosulfide # PubChem CID: 519823 IUPAC Name: 3-methylbutane-2-thiol SMILES: CC(C)C(C)S