CAS RN 209328-55-2
IUPAC Name:
5-fluoropyridin-3-ol
Synonyms:
5-fluoropyridin-3-ol
Molecular Weight (g/mol):
113.09
Molecular Formula:
C5H4FNO
InChi Key:
IUFLFKASIHPKNZ-UHFFFAOYSA-N
SMILES:
OC1=CC(F)=CN=C1
1
–
1
of
1
results
1
3-Fluoro-5-hydroxypyridine, 97%, Thermo Scientific™
CAS: 209328-55-2 Molecular Formula: C5H4FNO Molecular Weight (g/mol): 113.09 MDL Number: MFCD03095199 InChI Key: IUFLFKASIHPKNZ-UHFFFAOYSA-N Synonym: 3-fluoro-5-hydroxypyridine,5-fluoro-3-pyridinol,3-pyridinol, 5-fluoro,5-fluoro-3-hydroxypyridine,pubchem6625,acmc-209ffy,5-fluoranylpyridin-3-ol,ksc201g7t,3-pyridinol, 5-fluoro-9ci PubChem CID: 2762906 IUPAC Name: 5-fluoropyridin-3-ol SMILES: OC1=CC(F)=CN=C1