accessibility menu, dialog, popup

UserName

CAS RN 20938-79-8

H3CCH3HNOCH3OH

CAS RN 20938-79-8

IUPAC Name: 1-(3-methoxyphenyl)-2-[(propan-2-yl)amino]ethan-1-ol
Synonyms: 2-(isopropylamino)-1-(3-methoxyphenyl)ethanol
Molecular Weight (g/mol): 209.29
Molecular Formula: C12H19NO2
InChi Key: JZRMOFFUDLPSGK-UHFFFAOYNA-N
SMILES: COC1=CC=CC(=C1)C(O)CNC(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results