accessibility menu, dialog, popup

UserName

CAS RN 2096334-19-7

BOHNH3CFClHOOH

CAS RN 2096334-19-7

IUPAC Name: {2-chloro-5-[(2-fluoro-5-methylphenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-chloro-5-[(2-fluoro-5-methylphenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 307.51
Molecular Formula: C14H12BClFNO3
InChi Key: XLACLEABARDMPI-UHFFFAOYSA-N
SMILES: CC1=CC(NC(=O)C2=CC(B(O)O)=C(Cl)C=C2)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results