accessibility menu, dialog, popup

UserName

CAS RN 2096339-96-5

BONHCH3CH3FOHHO

CAS RN 2096339-96-5

IUPAC Name: {2-fluoro-5-[(2-methylpropyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-fluoro-5-[(2-methylpropyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 239.05
Molecular Formula: C11H15BFNO3
InChi Key: ZTGVLNJCMJKDDO-UHFFFAOYSA-N
SMILES: CC(C)CNC(=O)C1=CC(B(O)O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results