CAS RN 20972-36-5
IUPAC Name:
4-(4-methylphenyl)-4-oxobut-2-enoic acid
Synonyms:
4-(4-methylphenyl)-4-oxobut-2-enoic acid
Molecular Weight (g/mol):
190.2
Molecular Formula:
C11H10O3
InChi Key:
VNJMEFZKYZHWEO-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C(=O)C=CC(O)=O
1
–
1
of
1
results
1
3-(4-Methylbenzoyl)acrylic acid, 98%
CAS: 20972-36-5 Molecular Formula: C11H10O3 Molecular Weight (g/mol): 190.198 MDL Number: MFCD00075338 InChI Key: VNJMEFZKYZHWEO-VOTSOKGWSA-N Synonym: 3-4-methylbenzoyl acrylic acid,2e-4-4-methylphenyl-4-oxobut-2-enoic acid,e-4-oxo-4-p-tolyl but-2-enoic acid,trans-3-4-methylbenzoyl acrylic acid,e-3-p-toluoylacrylic acid,chembl44664,regid_for_cid_691106,3-4-methyl benzoyl acrylic acid,3-4-methyl-benzoyl acrylic acid,4-4-methylphenyl-4-oxo-2-butenoic acid PubChem CID: 691106 IUPAC Name: (E)-4-(4-methylphenyl)-4-oxobut-2-enoic acid SMILES: CC1=CC=C(C=C1)C(=O)C=CC(=O)O