accessibility menu, dialog, popup

UserName

CAS RN 209783-80-2

NH2NHONHOON

CAS RN 209783-80-2

IUPAC Name: (pyridin-3-yl)methyl N-({4-[(2-aminophenyl)carbamoyl]phenyl}methyl)carbamate
Synonyms: entinostat
Molecular Weight (g/mol): 376.42
Molecular Formula: C21H20N4O3
InChi Key: INVTYAOGFAGBOE-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1NC(=O)C1=CC=C(CNC(=O)OCC2=CC=CN=C2)C=C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (2)
  • (4)
  • (4)
  • (7)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (4)
  • (4)