CAS RN 2106-50-5
CAS RN 2106-50-5
IUPAC Name:
2-chloro-4-fluoro-1-nitrobenzene
Synonyms:
2-chloro-4-fluoro-1-nitrobenzene
Molecular Weight (g/mol):
175.54
Molecular Formula:
C6H3ClFNO2
InChi Key:
KQOOFMWRLDRDAX-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(F)C=C1Cl
1
–
2
of
2
results
1
2-Chloro-4-fluoro-1-nitrobenzene, 98%
CAS: 2106-50-5 Molecular Formula: C6H3ClFNO2 Molecular Weight (g/mol): 175.543 MDL Number: MFCD03412200 InChI Key: KQOOFMWRLDRDAX-UHFFFAOYSA-N Synonym: 2-chloro-4-fluoronitrobenzene,1-chloro-5-fluoro-2-nitrobenzene,benzene, 2-chloro-4-fluoro-1-nitro,pubchem4342,acmc-209fh8,2-choro-4-fluoronitrobenzene,ksc205m1p,3-chloro-4-nitrofluorobenzene,4-fluoro-2-chloronitrobenzene,2-chloro-4-fluoronitrobenzenen PubChem CID: 75017 IUPAC Name: 2-chloro-4-fluoro-1-nitrobenzene SMILES: C1=CC(=C(C=C1F)Cl)[N+](=O)[O-]
2
2-Chloro-4-fluoro-1-nitrobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.