CAS RN 212141-51-0
CAS RN 212141-51-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Vatalanib dihydrochloride, MedChemExpress
MedChemExpress Vatalanib dihydrochloride (PTK787 dihydrochloride) is an inhibitor of VEGFR2/KDR with IC50 of 37 nM.