accessibility menu, dialog, popup

UserName

CAS RN 21279-57-2

NHNNN

CAS RN 21279-57-2

IUPAC Name: 1-(pyrimidin-2-yl)-1,4-diazepane
Synonyms: 1-(pyrimidin-2-yl)-1,4-diazepane
Molecular Weight (g/mol): 178.24
Molecular Formula: C9H14N4
InChi Key: LZOGPVCTSDAYIP-UHFFFAOYSA-N
SMILES: C1CNCCN(C1)C1=NC=CC=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results