accessibility menu, dialog, popup

UserName

SOFFFOHSOFFFOHSOFFFOHH2OEr(E/Z)(E/Z)(E/Z)

CAS RN 21392-96-1

IUPAC Name: tris(1,1,1-trifluoro-4-hydroxy-4-(thiophen-2-yl)but-3-en-2-one) erbium hydrate
Synonyms: tris(1,1,1-trifluoro-4-hydroxy-4-(thiophen-2-yl)but-3-en-2-one) erbium hydrate
Molecular Weight (g/mol): 851.82
Molecular Formula: C24H17ErF9O7S3
InChi Key: LSMLVRQTTAIDBZ-UHFFFAOYSA-N
SMILES: O.[Er].OC(=CC(=O)C(F)(F)F)C1=CC=CS1.OC(=CC(=O)C(F)(F)F)C1=CC=CS1.OC(=CC(=O)C(F)(F)F)C1=CC=CS1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results