accessibility menu, dialog, popup

UserName

BrOH2NF

CAS RN 214210-17-0

IUPAC Name: 5-bromo-2-fluorobenzamide
Synonyms: 5-bromo-2-fluorobenzamide
Molecular Weight (g/mol): 218.03
Molecular Formula: C7H5BrFNO
InChi Key: MAGQVUXQRXLNGB-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results