accessibility menu, dialog, popup

UserName

CAS RN 21674-38-4

FFFFFFFFFFFFFFFNNNFFFFFFFFFFFFFFFFFFFFFFFFFFFFFF

CAS RN 21674-38-4

IUPAC Name: tris(1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptyl)-1,3,5-triazine
Synonyms: tris(1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptyl)-1,3,5-triazine
Molecular Weight (g/mol): 1185.21
Molecular Formula: C24F45N3
InChi Key: BFAUAMWYFFXWLX-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C1=NC(=NC(=N1)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)