accessibility menu, dialog, popup

UserName

CAS RN 216867-32-2

NSOHO

CAS RN 216867-32-2

IUPAC Name: 5-(pyridin-4-yl)thiophene-2-carboxylic acid
Synonyms: 5-(pyridin-4-yl)thiophene-2-carboxylic acid
Molecular Weight (g/mol): 205.23
Molecular Formula: C10H7NO2S
InChi Key: OJOWGMLSJKRJNV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(S1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results