CAS RN 216868-57-4
CAS RN 216868-57-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Deacetyl Linezolid Thioacetamide, TRC
CAS: 216868-57-4 Molecular Formula: C16 H20 F N3 O3 S Molecular Weight (g/mol): 353.41 Synonym: Linezolid Related Compound B (USP) IUPAC Name: N-[[(5S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]ethanethioamide SMILES: CC(=S)NC[C@H]1CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2
2
(S)-N-[[3-(3-Fluoro-4-morpholinophenyl)-2-oxooxazolidin-5-yl]methyl]thioacetamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.