accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHNHHCl

CAS RN 217806-26-3

IUPAC Name: tert-butyl N-(azetidin-3-yl)carbamate hydrochloride
Synonyms: tert-butyl N-(azetidin-3-yl)carbamate hydrochloride
Molecular Weight (g/mol): 208.69
Molecular Formula: C8H17ClN2O2
InChi Key: VLOLOLUAVFGUTD-UHFFFAOYSA-N
SMILES: Cl.CC(C)(C)OC(=O)NC1CNC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)