accessibility menu, dialog, popup

UserName

BrH3COOP(E)

CAS RN 2181-98-8

IUPAC Name: [(2E)-4-methoxy-4-oxobut-2-en-1-yl]triphenylphosphanium bromide
Synonyms: [(2E)-4-methoxy-4-oxobut-2-en-1-yl]triphenylphosphanium bromide
Molecular Weight (g/mol): 441.31
Molecular Formula: C23H22BrO2P
InChi Key: KSTXYAKHACCZSD-NWBUNABESA-M
SMILES: [Br-].COC(=O)\C=C\C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results