accessibility menu, dialog, popup

UserName

CAS RN 220239-66-7

CAS RN 220239-66-7

IUPAC Name: 3-(trifluoromethoxy)benzene-1-thiol
Synonyms: 3-(trifluoromethoxy)benzenethiol
Molecular Weight (g/mol): 194.17
Molecular Formula: C7H5F3OS
InChi Key: GEJGGOYNWFQKKH-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC(S)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results