accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CBOOCH3H3CCH3CH3(E/Z)

CAS RN 221006-76-4

IUPAC Name: 2-[1,2-diphenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[1,2-diphenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 432.17
Molecular Formula: C26H34B2O4
InChi Key: YFLBNMYBKXYNJN-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C(=C(B1OC(C)(C)C(C)(C)O1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results