CAS RN 22246-18-0
IUPAC Name:
7-hydroxy-1,2,3,4-tetrahydroquinolin-2-one
Synonyms:
7-hydroxy-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol):
163.18
Molecular Formula:
C9H9NO2
InChi Key:
LKLSFDWYIBUGNT-UHFFFAOYSA-N
SMILES:
OC1=CC2=C(CCC(=O)N2)C=C1
1
–
3
of
3
results
2
7-Hydroxy-3,4-dihydroquinolin-2(1H)-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.