accessibility menu, dialog, popup

UserName

CAS RN 2238-07-5

OOO

CAS RN 2238-07-5

IUPAC Name: 2-{[(oxiran-2-yl)methoxy]methyl}oxirane
Synonyms: diglycidyl ether
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: GYZLOYUZLJXAJU-UHFFFAOYNA-N
SMILES: C(OCC1CO1)C1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results