CAS RN 22876-20-6
CAS RN 22876-20-6
IUPAC Name:
6-chloro-2,3-dihydro-1,3-benzoxazole-2-thione
Synonyms:
6-chloro-3H-1,3-benzoxazole-2-thione
Molecular Weight (g/mol):
185.63
Molecular Formula:
C7H4ClNOS
InChi Key:
HAASPZUBSZGCKU-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2NC(=S)OC2=C1
No Refinements
1
–
0
of
0
results
1
6-Chloro-2-benzoxazolethiol, 99%, Thermo Scientific Chemicals
CAS: 22876-20-6 Molecular Formula: C7H4ClNOS Molecular Weight (g/mol): 185.63 MDL Number: MFCD01456482 InChI Key: HAASPZUBSZGCKU-UHFFFAOYSA-N Synonym: 6-chloro-1,3-benzoxazole-2-thiol,6-chloro-2-mercaptobenzoxazole,6-chloro-2-benzoxazolethiol,6-chlorobenzo d oxazole-2-thiol,2-mercapto-6-chlorobenzoxazole,6-chlorobenzoxazole-2 3h-thione,6-chlorobenzo d oxazole-2 3h-thione,6-chloro-benzooxazole-2-thiol,pubchem8714,6-chlorobenzoxazole-2-thiol PubChem CID: 2119214 IUPAC Name: 6-chloro-3H-1,3-benzoxazole-2-thione SMILES: C1=CC2=C(C=C1Cl)OC(=S)N2