CAS RN 22907-72-8
IUPAC Name:
1,4-bis(propan-2-yl)cyclohexane
Synonyms:
1,4-diisopropylcyclohexane
Molecular Weight (g/mol):
168.32
Molecular Formula:
C12H24
InChi Key:
GMUJGUXGVJZHSK-UHFFFAOYSA-N
SMILES:
CC(C)C1CCC(CC1)C(C)C
1
–
1
of
1
results
1
1,4-Diisopropylcyclohexane, cis + trans, 98%, Thermo Scientific™
CAS: 22907-72-8 Molecular Formula: C12H24 Molecular Weight (g/mol): 168.324 MDL Number: MFCD00019366 InChI Key: GMUJGUXGVJZHSK-UHFFFAOYSA-N Synonym: 1,4-diisopropylcyclohexane,1,4-diisopropyl cyclohexane,1,4-di-isopropylcyclohexane,1,4-di propan-2-yl cyclohexane,cyclohexane,1,4-bis 1-methylethyl-,trans,acmc-1cbxy,1,4-diisopr opylcyclohexane,1,4-bis methylethyl cyclohexane,1alpha,4beta-diisopropylcyclohexane,1,4-diisopropylcyclohexane,cis+trans PubChem CID: 140972 IUPAC Name: 1,4-di(propan-2-yl)cyclohexane SMILES: CC(C)C1CCC(CC1)C(C)C