CAS RN 229971-81-7
CAS RN 229971-81-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
MedChemExpress ADH-1
MedChemExpress ADH-1, an N-cadherin antagonist, inhibits N-cadherin mediated cell adhesion.
2
MedChemExpress ADH-1
MedChemExpress ADH-1, an N-cadherin antagonist, inhibits N-cadherin mediated cell adhesion.
3
MedChemExpress ADH-1
MedChemExpress ADH-1, an N-cadherin antagonist, inhibits N-cadherin mediated cell adhesion.