CAS RN 23039-94-3
IUPAC Name:
tris(3-fluorophenyl)phosphane
Synonyms:
tris(3-fluorophenyl)phosphane
Molecular Weight (g/mol):
316.26
Molecular Formula:
C18H12F3P
InChi Key:
CUTRINLXFPIWQB-UHFFFAOYSA-N
SMILES:
FC1=CC(=CC=C1)P(C1=CC(F)=CC=C1)C1=CC=CC(F)=C1
1
–
1
of
1
results
1
Tris(3-fluorophenyl)phosphine, 97%
CAS: 23039-94-3 Molecular Formula: C18H12F3P Molecular Weight (g/mol): 316.263 MDL Number: MFCD00015722 InChI Key: CUTRINLXFPIWQB-UHFFFAOYSA-N Synonym: tris 3-fluorophenyl phosphine,tris 3-fluorophenyl phosphane,tri 3-fluorophenyl phosphine,phosphine,tris 3-fluorophenyl,acmc-1cqwl,tri m-fluorophenyl phosphine,tri-3-fluorophenyl-phosphine PubChem CID: 140988 IUPAC Name: tris(3-fluorophenyl)phosphane SMILES: C1=CC(=CC(=C1)P(C2=CC=CC(=C2)F)C3=CC=CC(=C3)F)F