CAS RN 2308035-51-8
CAS RN 2308035-51-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thalidomide-O-amido-C3-COOH, MedChemExpress
MedChemExpress Thalidomide-O-amido-C3-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.