CAS RN 2311-46-8
IUPAC Name:
propan-2-yl hexanoate
Synonyms:
isopropyl hexanoate
Molecular Weight (g/mol):
158.24
Molecular Formula:
C9H18O2
InChi Key:
JSHDAORXSNJOBA-UHFFFAOYSA-N
SMILES:
CCCCCC(=O)OC(C)C
1
–
3
of
3
results
1
N-Caproic Acid Isopropyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Caproic Acid Isopropyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Caproic Acid Isopropyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.