accessibility menu, dialog, popup

UserName

ClFFFCl

CAS RN 23131-73-9

IUPAC Name: 1-chloro-4-(chloromethyl)-2-(trifluoromethyl)benzene
Synonyms: 1-chloro-4-(chloromethyl)-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 229.02
Molecular Formula: C8H5Cl2F3
InChi Key: NREUBTUYADXMHQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C=CC(CCl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results