CAS RN 23554-99-6
CAS RN 23554-99-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress HOE 32020
MedChemExpress HOE 32020 is a Hoechst stain, which is a blue fluorescent dyes used to stain DNA.