CAS RN 2360-22-7
CAS RN 2360-22-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methyl-1H-benzimidazole-2-thiol, TRC
CAS: 2360-22-7 Chemical Name or Material: 1-Methyl-1H-benzimidazole-2-thiol InChI Formula: InChI=1S/C8H8N2S/c1-10-7-5-3-2-4-6(7)9-8(10)11/h2-5H,1H3,(H,9,11) IUPAC Name: 1-methylbenzimidazole-2-thiol Molecular Formula: C8H8N2S Molecular Weight (g/mol): 164.22