accessibility menu, dialog, popup

UserName

CAS RN 2373-79-7

IUPAC Name: (2E)-3-(4-ethoxyphenyl)prop-2-enoate
Synonyms: (2E)-3-(4-ethoxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 191.21
Molecular Formula: C11H11O3
InChi Key: DZLOUWYGNATKKZ-VMPITWQZSA-M
SMILES: CCOC1=CC=C(\C=C\C([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results