CAS RN 2384108-14-7
CAS RN 2384108-14-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Pivalamido Cefazolin, TRC
CAS: 2384108-14-7 Molecular Formula: C16H20N4O4S3 Molecular Weight (g/mol): 428.55 Synonym: Cefazolin EP Impurity B,(6R,7R)-3-(((5-Methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-7-pivalamido-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid IUPAC Name: (6R,7R)-7-(2,2-dimethylpropanoylamino)-3-[(5-methyl-1,3,4-thiadiazol-2-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid SMILES: Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](NC(=O)C(C)(C)C)C3=O)C(=O)O)s1