accessibility menu, dialog, popup

UserName

CAS RN 2387-23-7

OHNHN

CAS RN 2387-23-7

IUPAC Name: 1,3-dicyclohexylurea
Synonyms: dicyclohexylurea
Molecular Weight (g/mol): 224.35
Molecular Formula: C13H24N2O
InChi Key: ADFXKUOMJKEIND-UHFFFAOYSA-N
SMILES: O=C(NC1CCCCC1)NC1CCCCC1
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)