accessibility menu, dialog, popup

UserName

OHFFFF

CAS RN 238742-82-0

IUPAC Name: [5-fluoro-2-(trifluoromethyl)phenyl]methanol
Synonyms: [5-fluoro-2-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 194.13
Molecular Formula: C8H6F4O
InChi Key: AGVUJWYLCSOJDS-UHFFFAOYSA-N
SMILES: OCC1=CC(F)=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results