accessibility menu, dialog, popup

UserName

CAS RN 2396-65-8

HCCH

CAS RN 2396-65-8

IUPAC Name: nona-1,8-diyne
Synonyms: 1,8-nonadiyne
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: DMOVPHYFYSASTC-UHFFFAOYSA-N
SMILES: C#CCCCCCC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)