accessibility menu, dialog, popup

UserName

CAS RN 2403-89-6

CH3NCH3CH3HOH3CCH3

CAS RN 2403-89-6

IUPAC Name: 1,2,2,6,6-pentamethylpiperidin-4-ol
Synonyms: 1,2,2,6,6-pentamethylpiperidin-4-ol
Molecular Weight (g/mol): 171.28
Molecular Formula: C10H21NO
InChi Key: NWHNXXMYEICZAT-UHFFFAOYSA-N
SMILES: CN1C(C)(C)CC(O)CC1(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results